Learn More
StatLab PathFlow PiSmart Thermal Transfer Attached Lid Front Hinge Biopsy Cassettes
SDP
Catalog No. 21200168
Click to view available options
Color:
Blue
Gray
Green
Lavender
Orange
Pink
Tan
White
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy
Thermal transfer biopsy cassettes with attached lid and front hinge.
  • 0.9 mm pores
  • Front hinge with lid attached
  • Made from acetal polymer
  • Specifically validated for the Pi Smart Thermal Transfer Cassette Printer
  • Print face features four raised pips in the corners to protect printed surface during dewaxing, while enabling the surface to be cleared of wax
  • Threaded
  • 2000 per case
Specifications
Quantity 2000 Cassettes/Cs.
Color Blue
Product Type Biopsy Cassettes
Made in USA
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.