Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ PathoDx Strep Grouping Universal Kit

Catalog No. p-8401010
Change view
Click to view available options
Quantity:
60 Tests/Kit
Catalog No. Quantity
R62076 60 Tests/Kit
1 options
This item is not returnable. View return policy

Catalog No. R62076

Supplier: Thermo Scientific™ R62076

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Support the functionality of the Thermo Scientific™ PathoDx™ Strep Grouping Kit with Thermo Scientific™ PathoDx™ Strep Grouping Universal Kit.

Support the functionality of the Thermo Scientific™ PathoDx™ Strep Grouping Kit with Thermo Scientific™ PathoDx™ Strep Grouping Universal Kit. This kit is a set of components to be used with selected, separately purchased PathoDxStrep Grouping Reagents designed to identify beta-haemolytic streptococci of Lancefield Groups A, B, C, F or G from primary culture plates.This accessory kit contains: Extraction reagent 1; Extraction reagent 2; Extraction reagent 3; Positive control; Reaction cards; Stirrers.

Includes:

Control antigens ABC and FG; extraction reagents 1, 2, 3; stirrers; disposable slides

Requires:

For use with Part No. R62030, R62031, R62032, R62034, R62035.

TRUSTED_SUSTAINABILITY

Specifications

Control Sets Positive
Description Strep Universal Grouping Kit
Detectable Analytes beta-hemolytic streptococci (Groups A, B, C, F, G)
Type Streptococcus Test Kit
Includes Control antigens ABC and FG; extraction reagents 1, 2, 3; stirrers; disposable slides
Organism Group Lancefield A, B, C, D, F and G
Quantity 60 Tests/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.