Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PATJ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585444
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585444 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585444 Supplier Invitrogen™ Supplier No. PA585444
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (84%), Rat (85%), Bovine (88%). Positive Control: RT-4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein with multiple PDZ domains. PDZ domains mediate protein-protein interactions, and proteins with multiple PDZ domains often organize multimeric complexes at the plasma membrane. This protein localizes to tight junctions and to the apical membrane of epithelial cells. A similar protein in Drosophila is a scaffolding protein which tethers several members of a multimeric signaling complex in photoreceptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PATJ
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene PATJ
Gene Accession No. Q8NI35
Gene Alias channel-interacting PDZ domain protein; channel-interacting PDZ domain-containing protein; Cipp; hINADL; inactivation no after-potential D-like protein; inactivation-no-afterpotential D-like; Inadl; Inadl protein; Inadl2; InaD-like; InaD-like (Drosophila); InaD-like 2; inaD-like protein; novel ankyrin-repeat protein; PALS1-associated tight junction protein; Patj; PATJ, crumbs cell polarity complex component; PDZ domain protein; protein associated to tight junctions; RGD1565362
Gene Symbols PATJ
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human PATJ. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10207, 140581
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.