Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PATJ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597072
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597072 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597072 Supplier Invitrogen™ Supplier No. PA597072
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: SKOV3, SW480, 293T, HeLa; Cellular Location: Apical cell membrane, Cell junction, Cell membrane, Cytoplasm, Peripheral membrane protein, perinuclear region, tight junction Immunogen sequence: VRRKTSSSTS PLEPPSDRGT VVEPLKPPAL FLTGAVETET NVDGEDEEIK ERIDTLKNDN IQALEKLEKV PDSPENELKS RWENLLGPDY EVMVATLDTQ IADDAELQKY SKLLPIHTLR LGVEVDSFDG HHYISSIVSG GPVDTLGLLQ PEDELLEVNG MQLYGKSRRE AVSFLKEVPP PFTLVCCRRL FDDEASVDEP RRTETSLPET EVDHNMDVNT EEDDDGELAL W.

This gene encodes a protein with multiple PDZ domains. PDZ domains mediate protein-protein interactions, and proteins with multiple PDZ domains often organize multimeric complexes at the plasma membrane. This protein localizes to tight junctions and to the apical membrane of epithelial cells. A similar protein in Drosophila is a scaffolding protein which tethers several members of a multimeric signaling complex in photoreceptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PATJ
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene PATJ
Gene Accession No. Q63ZW7, Q8NI35
Gene Alias channel-interacting PDZ domain protein; channel-interacting PDZ domain-containing protein; Cipp; hINADL; inactivation no after-potential D-like protein; inactivation-no-afterpotential D-like; Inadl; Inadl protein; Inadl2; InaD-like; InaD-like (Drosophila); InaD-like 2; inaD-like protein; novel ankyrin-repeat protein; PALS1-associated tight junction protein; Patj; PATJ, crumbs cell polarity complex component; PDZ domain protein; protein associated to tight junctions; RGD1565362
Gene Symbols PATJ
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 450-680 of human INADL (NP_7953522).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10207, 12695
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.