Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PATL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589047
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589047 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589047 Supplier Thermo Scientific Supplier No. PA589047
Add to Cart
Add to Cart

PATL1 Polyclonal Antibody

PATL1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen PATL1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Patl1
Gene Accession No. B5DF93, Q3TC46
Gene Alias AV312086; hPat1b; OK/KNS-cl.5; PAT1 homolog 1, processing body mRNA decay factor; Pat1b; PAT1-like protein 1; PATL1; protein associated with topoisomerase II homolog 1; protein associated with topoisomerase II homolog 1 (yeast); protein PAT1 homolog 1; Protein PAT1 homolog b; RGD1305514
Gene Symbols Patl1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 535-640 of human PATL1 (NP_6899292).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225929, 361736
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.