Learn More
Invitrogen™ PATZ1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total PATZ1 protein.
Specifications
Specifications
| Antigen | PATZ1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 2.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | PATZ1 |
| Gene Accession No. | Q9HBE1 |
| Gene Alias | 8430401L15Rik; AI447519; BTB/POZ domain zinc finger transcription factor; BTB-POZ domain zinc finger transcription factor; dJ400N23; Mazr; MAZ-related factor; Patz; PATZ1; POZ (BTB) and AT hook containing zinc finger 1; POZ-, AT hook-, and zinc finger-containing protein 1; POZ/BTB and AT hook containing zinc finger 1; POZ-AT hook-zinc finger protein; protein kinase A RI subunit alpha-associated protein; RIAZ; transcription factor (MAZ-related) MAZR; ZBTB19; Zfp278; zi; Zinc finger and BTB domain-containing protein 19; zinc finger protein 278; Zinc finger sarcoma gene protein; ZNF278; ZSG |
| Gene Symbols | PATZ1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.