Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PauA Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989392
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9392 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9392 Supplier Bioss Supplier No. BS9846R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Plasminogen activator A (PauA) is a Streptococcus uberis protein that functions similarly to proteins within the Staphylokinase/Streptokinase family. Streptococcus uberis is a common cause of infection and inflammatory disease in the bovine udder. Inflammation of breast tissue (mastitis) has been recognized as a serious threat to the welfare of the dairy cow and has a severe economic impact on the dairy industry. S. uberis has been shown to specifically activate bovine and ovine plasminogen and this is mediated by the extracellular protein, plasminogen activator uberis (PauA), which has a molecular mass of around 30 kDa. It has been postulated that activation of plasminogen by PauA may facilitate early bacterial colonization of the lactating gland by promoting the release of nutrients which are likely to be restricted in milk. (https://www.ncbi.nlm.nih.gov/pubmed/10483719).

Specifications

Antigen PauA
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from Streptococcus uberis PauA protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.