Learn More
PAX-8, Mouse anti-Human, Clone: 15C8.1, Millipore Sigma™
Mouse Monoclonal Antibody
Supplier:  MilliporeSigma MABS189925UG
Catalog No. MABS189925
Only null left
Add to cart
Description
Description
Anti-PAX-8, clone 15C8.1, Cat. No. MABS1899, is a mouse monoclonal antibody for detection of human Paired box protein Pax-8 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Paired box protein Pax-8 (UniProt: Q06710; also known as PAX-8) is encoded by the Pax8 gene (Gene ID: 7849) in human. PAX-8 is a member of the paired box (PAX) family of transcription factors. PAX proteins are important regulators of organogenesis and act as key factors in maintaining pluripotency of stem cells during development. PAX-8 acts as a transcription factor for the thyroid-specific expression of the genes and is exclusively expressed in the thyroid cell type, maintaining the functional differentiation of such cells. PAX-8 is shown to be crucial in determining cell fate during the development of the thyroid, kidney, brain, eyes, and Müllerian system. It also regulates the expression of the Wilms' tumor suppressor gene (WT1). PAX-8 is a shown to be a highly sensitive marker for thyroid, renal, Müllerian, and thymic tumors. However, lung adenocarcinomas, breast and adrenal neoplasms, and the majority of gastrointestinal tumors are reported to be negative for PAX-8. Mutations in Pax8 gene are known to cause congenital hypothyroidism that is characterized by thyroid dysgenesis and in some cases the thyroid gland can be completely absent, ectopically located and/or severely hypoplastic. Five isoforms of PAX-8 have been described that are produced by alternative splicing. (Ref.: Laury, AR et al. (2011). Am J Surg. Pathol. 35(6); 816-826; Xiang, L and Kong, B (2013). Oncol. Lett. 5(3); 735-738).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PAX-8
Monoclonal
Unconjugated
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Pax8
GST/His-tagged recombinant fragment corresponding to 129 amino acids from the internal region of human Pax-8.
25 μg
Signaling
Clone 15C8.1 detects Pax-8 protein in human ovarian cells. It targets an epitope within 129 amino acids from the internal region.
Stable for 1 year at 2-8°C from date of receipt.
IgG1
Immunohistochemistry (Paraffin), Western Blot
15C8.1
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Pax-8 in human uterine tube tissue sections.
Paired box protein Pax-8
Mouse
Protein G purified
RUO
Primary
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.