Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551033
Description
PAX1 Polyclonal Antibody detects endogenous levels of PAX1 protein.
The PAX genes, including PAX1, are a highly conserved family of developmental control genes that encode transcription factors and have been shown to play a role in pattern formation during embryogenesis in vertebrates. See PAX7 (MIM 167410) for a discussion of paired box domain genes.
Specifications
PAX1 | |
Polyclonal | |
Unconjugated | |
PAX1 | |
hbs; hunchback; HuP48; OFC2; paired box 1; paired box gene 1; Paired box homeotic gene 1; paired box protein Pax-1; paired domain gene HuP48; PAX1; Pax-1; un; undulated; wavy tail; wt | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5075 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P15863 | |
PAX1 | |
Synthesized peptide derived from the Internal region of human Pax-1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction