Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax2 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299013JF549
Description
Pax2 Polyclonal antibody specifically detects Pax2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
Pax2 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
paired box 2, paired box gene 2, paired box homeotic gene 2, paired box protein Pax-2 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Pax2. | |
0.1 mL | |
Chaperone Mediated Autophagy (CMA), Neuronal Cell Markers, Stem Cell Markers | |
5076 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction