Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax2 Antibody (PAX2/1104), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279877AF594
Description
Pax2 Monoclonal specifically detects Pax2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Pax2 | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PAX2 | |
Recombinant fragment (around aa 223-354) human Pax2 protein (exact sequence is proprietary) (Uniprot: Q02962) | |
0.1 mL | |
Chaperone Mediated Autophagy (CMA), Neuronal Cell Markers, Stem Cell Markers | |
This antibody recognizes a protein of 42kDa, which is identified as Pax2. Pax2 has been used as a marker for the identification of renal cell carcinoma and ovarian carcinoma by immunohistochemistry. | |
Store at 4°C in the dark. |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
PAX2/1104 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
paired box 2, paired box gene 2, paired box homeotic gene 2, paired box protein Pax-2 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction