Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAX2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572630
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572630 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572630 Supplier Invitrogen™ Supplier No. PA572630
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PAX2A Polyclonal Antibody for Western Blot

Probable transcription factor involved in the development of the midbrain/hindbrain boundary (MHB) organizer and specification of neuronal cell fates. Required for establishment of eng2 and eng3 gene expression in the midbrain and MHB primordium during late gastrulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAX2A
Applications Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene pax2a
Gene Accession No. Q90268
Gene Alias cb378; No isthmus protein; noi; paired box 2a; paired box gene 2.1; paired box gene 2a; paired box homeotic gene 2a; paired box protein pax2.1 splice variant; paired box protein Pax-2a; pax[zf-b]; Pax-2; pax2.1; pax21; pax2a; pax2a1; paxb; pax-b; PAXZF-B
Gene Symbols pax2a
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 258-284 amino acids from the Central region of DANRE pax2a.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30425
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.