Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax3 Antibody (GT2411), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP313612
This item is not returnable.
View return policy
Description
Pax3 Monoclonal antibody specifically detects Pax3 in Human samples. It is validated for Western BlotSpecifications
| Pax3 | |
| Monoclonal | |
| Unconjugated | |
| CDHS, HuP2, HUP2MGC120384, MGC120381, MGC120382, MGC120383, MGC134778, paired box 3, paired box gene 3 (Waardenburg syndrome 1), paired box homeotic gene 3, paired box protein Pax-3, paired domain gene 3, paired domain gene HuP2, Waardenburg syndrome 1, WS1, WS3 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Pax3. The exact sequence is proprietary. | |
| 100 μg | |
| Apoptosis, Mesenchymal Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators | |
| 5077 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot | |
| GT2411 | |
| PBS, 20% Glycerol (pH7) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction