Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax3 Antibody (PAX3/4700), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP320576100UG
Description
Pax3 Monoclonal antibody specifically detects Pax3 in Human, Avian samples. It is validated for ELISASpecifications
Pax3 | |
Monoclonal | |
0.2 mg/ml | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Avian | |
Purified |
ELISA | |
PAX3/4700 | |
Unconjugated | |
CDHS, HuP2, HUP2MGC120384, MGC120381, MGC120382, MGC120383, MGC134778, paired box 3, paired box gene 3 (Waardenburg syndrome 1), paired box homeotic gene 3, paired box protein Pax-3, paired domain gene 3, paired domain gene HuP2, Waardenburg syndrome 1, WS1, WS3 | |
Synthetic peptide (quail) from the C-terminus | |
100 μg | |
Apoptosis, Mesenchymal Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators | |
5077 | |
Store at 4&drg;C. Do not freeze. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction