Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAX3 Monoclonal Antibody (GT2411)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531582
Description
Keep as concentrated solution. Positive Control: U87-MG, U87-MG nuclear extract. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Pax 3 (Paired Box 3) is a transcription factor that is involved in development of the peripheral nervous system, melanocytes, some vascular smooth muscle, and a number of other derivatives. It regulates neurogenesis in pre-migratory neural crest cells from the dorsal neural tube, and in myogenic progenitors in the presomitic mesoderm and the hypaxial somites. Alternative splicing results in isoforms having different C-termini.
Specifications
PAX3 | |
Monoclonal | |
1 mg/mL | |
PBS with 20% glycerol and no preservative; pH 7 | |
P23760, P24610 | |
PAX3 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Human PAX3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2a |
Western Blot | |
GT2411 | |
Unconjugated | |
PAX3 | |
CDHS; homeodomain protein PAX3; HUP2; MGC120381; MGC120382; MGC120383; MGC120384; MGC134778; paired box 3; paired box gene 3; paired box homeotic gene 3; paired box protein Pax-3; paired domain gene 3; paired domain gene HuP2; PAX3; Pax-3; PAX3/FKHR fusion; Sp; splotch; WS1; WS3 | |
Mouse | |
Protein A | |
RUO | |
114502, 18505, 5077 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction