Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAX3 Mouse anti-Human, Mouse, Clone:7D8G7, Abnova™
Mouse monoclonal antibody raised against recombinant human PAX3.
Supplier: Abnova Corporation MAB17812
Description
This gene is a member of the paired box (PAX) family of transcription factors. Members of the PAX family typically contain a paired box domain and a paired-type homeodomain. These genes play critical roles during fetal development. Mutations in paired box gene 3 are associated with Waardenburg syndrome, craniofacial-deafness-hand syndrome, and alveolar rhabdomyosarcoma. The translocation t(2;13)(q35;q14), which represents a fusion between PAX3 and the forkhead gene, is a frequent finding in alveolar rhabdomyosarcoma. Alternative splicing results in transcripts encoding isoforms with different C-termini. [provided by RefSeq]Specifications
PAX3 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Flow Cytometry Immunocytochemistry Immunohistochemistry Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. |
|
PAX3 | |
Mouse | |
100 μg | |
Human, Mouse | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
7D8G7 | |
paired box 3 | |
In PBS (0.05% sodium azide) | |
CDHS, HUP2, MGC120381, MGC120382, MGC120383, MGC120384, MGC134778, WS1 | |
Recombinant protein corresponding to amino acids 142-203 of human PAX3 from E. coli. | |
5077 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction