Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax3/Pax7 Antibody (274212), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2457I
Description
Pax3/Pax7 Monoclonal antibody specifically detects Pax3/Pax7 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOFSpecifications
Pax3/Pax7 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
E. coli-derived recombinant human Pax3 (isoform Pax3a), Met1-Ser215, Accession # NP_000429 | |
0.1 mL | |
Apoptosis, Mesenchymal Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
274212 | |
Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction