Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAX3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981010
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1010 Supplier Bioss Supplier No. BS1097R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Pax 3 (Paired Box 3) is a transcription factor that is involved in development of the peripheral nervous system, melanocytes, some vascular smooth muscle, and a number of other derivatives. It regulates neurogenesis in pre-migratory neural crest cells from the dorsal neural tube, and in myogenic progenitors in the presomitic mesoderm and the hypaxial somites. Alternative splicing results in isoforms having different C-termini.

Specifications

Antigen PAX3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PAX3
Gene Accession No. P23760
Gene Alias CDHS; homeodomain protein PAX3; HUP2; MGC120381; MGC120382; MGC120383; MGC120384; MGC134778; paired box 3; paired box gene 3; paired box homeotic gene 3; Paired box protein Pax-3; paired domain gene 3; paired domain gene HuP2; Pax3; Pax-3; PAX3/FKHR fusion; Sp; splotch; WS1; WS3
Gene Symbols PAX3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse PAX3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5077
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.