Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAX3 Recombinant Rabbit Monoclonal Antibody (HL160)
GREENER_CHOICE

Catalog No. PIMA538267
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538267 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538267 Supplier Invitrogen™ Supplier No. MA538267
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Centrifuge briefly prior to opening.

Pax 3 (Paired Box 3) is a transcription factor that is involved in development of the peripheral nervous system, melanocytes, some vascular smooth muscle, and a number of other derivatives. It regulates neurogenesis in pre-migratory neural crest cells from the dorsal neural tube, and in myogenic progenitors in the presomitic mesoderm and the hypaxial somites. Alternative splicing results in isoforms having different C-termini.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAX3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL160
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PAX3
Gene Accession No. P23760, P24610
Gene Alias CDHS; homeodomain protein PAX3; HUP2; MGC120381; MGC120382; MGC120383; MGC120384; MGC134778; paired box 3; paired box gene 3; paired box homeotic gene 3; paired box protein Pax-3; paired domain gene 3; paired domain gene HuP2; PAX3; Pax-3; PAX3/FKHR fusion; Sp; splotch; WS1; WS3
Gene Symbols PAX3
Host Species Rabbit
Immunogen The immunogen used to generate this antibody corresponds to Coturnix coturnix PAX3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18505, 5077
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.