Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAX4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596316
Description
Immunogen sequence: LGRYYRTGVL EPKGIGGSKP RLATPPVVAR IAQLKGECPA LFAWEIQRQL CAEGLCTQDK TPSVSSINRV LRALQEDQGL PCTRLRSPAV LAPAVLTPHS GSETPRGTHP GTGHRNRTIF SPSQAEALEK EFQRGQYPDS VARGKLATAT S; Positive Samples: Lovo; Cellular Location: Nucleus.
This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The paired box 4 gene is involved in pancreatic islet development and mouse studies have demonstrated a role for this gene in differentiation of insulin-producing beta cells.
Specifications
PAX4 | |
Polyclonal | |
Unconjugated | |
PAX4 | |
KPD; MGC129960; MODY9; paired box 4; paired box gene 4; paired box protein Pax-4; paired domain gene 4; Pax4; Pax-4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
18506, 5078 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.52 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43316, P32115 | |
PAX4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 50-200 of human PAX4 (NP_0061842). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction