Learn More
Pax5/BSAP Antibody (1207C), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB3487U
Description
Pax5/BSAP Monoclonal antibody specifically detects Pax5/BSAP in Human, Mouse samples. It is validated for Western Blot, Western Blot, Flow Cytometry, Immunohistochemistry, ImmunocytochemistrySpecifications
Pax5/BSAP | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
1207C | |
Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5 | |
E. coli-derived recombinant human Pax5, Thr141-His391, Accession # Q02548 | |
0.1 mL | |
B Cell Development and Differentiation Markers, Cancer, Transcription Factors and Regulators | |
5079 | |
Store at 4C in the dark. | |