Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax5/BSAP Antibody (1207C) [PerCP], Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB3487C
Description
Pax5/BSAP Monoclonal antibody specifically detects Pax5/BSAP in Human, Mouse samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, ImmunocytochemistrySpecifications
Pax5/BSAP | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
1207C | |
Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5 | |
E. coli-derived recombinant human Pax5, Thr141-His391, Accession # Q02548 | |
0.1 mL | |
B Cell Development and Differentiation Markers, Cancer, Transcription Factors and Regulators | |
5079 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction