Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax5/BSAP Antibody (1H9), mFluor Violet 610 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP200258MFV610
Description
Pax5/BSAP Monoclonal antibody specifically detects Pax5/BSAP in Human, Mouse samples. It is validated for Flow CytometrySpecifications
Pax5/BSAP | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry | |
1H9 | |
Flow Cytometry | |
B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5 | |
The immunogen for this antibody is Pax5, BSAP (1H9). | |
0.1 mL | |
B Cell Development and Differentiation Markers, Cancer, Transcription Factors and Regulators | |
5079 | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction