Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax5/BSAP Antibody (PAX5/3977R), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308310R
Description
Pax5/BSAP Monoclonal antibody specifically detects Pax5/BSAP in Human, Canine samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Pax5/BSAP | |
Monoclonal | |
DyLight 550 | |
B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5 | |
A recombinant fragment (around aa 235-382) of human PAX5 protein (exact sequence is proprietary) (Uniprot: Q02548) | |
100 μg | |
B Cell Development and Differentiation Markers, Cancer, Transcription Factors and Regulators | |
5079 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PAX5/3977R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction