Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax5/BSAP Mouse anti-Human, Clone: rPAX5/2060, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | Pax5/BSAP |
---|---|
Clone | rPAX5/2060 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, ELISA |
Applications | Western Blot, ELISA |
Description
Pax5/BSAP Monoclonal specifically detects Pax5/BSAP in Human samples. It is validated for Western Blot, ELISA.Specifications
Pax5/BSAP | |
0.2 mg/ml | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
B Cell Development and Differentiation Markers, Cancer, Transcription Factors and Regulators | |
10 mM PBS with 0.05% BSA | |
5079 | |
IgG1 κ | |
Protein A or G purified |
rPAX5/2060 | |
Western Blot 1-2 ug/ml, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5 | |
Recombinant full-length humanPax5/BSAP protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title