Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAX5 Monoclonal Antibody (7D3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548566
Description
PAX5 Monoclonal Antibody for Western Blot, ICC/IF, IHC, Flow, ELISA
PAX5 is a member of the paired box (PAX) family of transcription factors. The PAX proteins are important regulators in early development and alterations in the expression of their genes are thought to contribute to neoplastic transformation. PAX5 is the B-cell lineage specific activator protein (BSAP) that is expressed at early, but not late stages, of B-cell differentiation. Its expression has also been detected in developing CNS and testis, therefore, PAX5 may not only play an important role in B-cell differentiation, but also in neural development and spermatogenesis. Mutations in the gene can result in leukemia, acute lymphoblastic 3.
Specifications
PAX5 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q02548 | |
PAX5 | |
Purified recombinant fragment of human PAX5 (AA: 235-382) expressed in E. Coli. ; ;. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry | |
7D3 | |
Unconjugated | |
PAX5 | |
ALL3; B-cell lineage specific activator; B-cell-specific transcription factor; BSAP; EBB-1; KLP; paired box 5; paired box gene 5; paired box gene 5 (B-cell lineage specific activator); paired box homeotic gene 5; paired box protein Pax-5; paired domain gene 5; Pax5; Pax-5; Pax5 variant 1; Pax5 variant 2; RGD1565236; transcription factor PAX 5 | |
Mouse | |
Protein G | |
RUO | |
5079 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction