Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAX5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA1109
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA1109 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA1109 Supplier Invitrogen™ Supplier No. PA1109
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Western blot analysis of PA1-109 detects a prominent ~45 kDa protein in PAX5 expressing human Ramos, Daudi, Gaji and mouse A20 B-cell lymphoma cells. No PAX5 was detected in negative control Jurkat cells. The 45 kDa band is enriched in immunoprecipitations from Raji lysate, however additional unknown high molecular weight bands are also enriched and detected by PA1-109. PAX5 was also specifically detected by immunofluorescence in the nuclei of Ramos cells compared to negative control Jurkat cells.

PAX5 is a member of the paired box (PAX) family of transcription factors. The PAX proteins are important regulators in early development and alterations in the expression of their genes are thought to contribute to neoplastic transformation. PAX5 is the B-cell lineage specific activator protein (BSAP) that is expressed at early, but not late stages, of B-cell differentiation. Its expression has also been detected in developing CNS and testis, therefore, PAX5 may not only play an important role in B-cell differentiation, but also in neural development and spermatogenesis. Mutations in the gene can result in leukemia, acute lymphoblastic 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAX5
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gene PAX5
Gene Accession No. Q02548, Q02650
Gene Alias ALL3; B-cell lineage specific activator; B-cell-specific transcription factor; BSAP; EBB-1; KLP; paired box 5; paired box gene 5; paired box gene 5 (B-cell lineage specific activator); paired box homeotic gene 5; paired box protein Pax-5; paired domain gene 5; Pax5; Pax-5; Pax5 variant 1; Pax5 variant 2; RGD1565236; transcription factor PAX 5
Gene Symbols PAX5
Host Species Rabbit
Immunogen Purified internal fragment of human recombinant PAX5 expressed in E. coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18507, 5079
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.