Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAX5 Rabbit anti-Human, Unconjugated, Clone: RM331, Abnova™

Catalog No. 89339191 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-191 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-191 Supplier Abnova Corporation Supplier No. MAB21974
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

PAX5 Monoclonal antibody specifically detects PAX5 in Human samples. It is validated for WB-Ce, IHC-P.

Specifications

Antigen PAX5
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM331
Conjugate Unconjugated
Description paired box 5
Gene PAX5
Gene Alias BSAP
Host Species Rabbit
Immunogen A peptide corresponding to C-terminus of human PAX5
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 5079
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.