Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax6 Antibody (22C3), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Pax6 |
---|---|
Clone | 22C3 |
Dilution | Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 |
Applications | Flow Cytometry, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
Pax6 Monoclonal antibody specifically detects Pax6 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
Pax6 | |
Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6 | |
A synthesized peptide derived from Human Pax6 [UniProt P26367] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
22C3 | |
Flow Cytometry, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cellular Markers, Diabetes Research, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
5080 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title