Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax6 Antibody (PAX6/498), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234705B
Description
Pax6 Monoclonal specifically detects Pax6 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).Specifications
Pax6 | |
Monoclonal | |
Biotin | |
keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6 | |
Mouse | |
47 kDa | |
0.1 mL | |
Cellular Markers, Diabetes Research, Neuronal Stem Cell Markers, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
5080 | |
Human, Mouse, Rat, Chicken | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
PAX6/498 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
PAX6 | |
Recombinant fragment (N-terminus; aa 1-300) of human PAX6 protein | |
Protein A or G purified | |
RUO | |
Primary | |
Pax genes contain paired domains with strong homology to genes in Drosophila, which are involved in programming early development. Lesions in the Pax-6 gene account for most cases of aniridia, a congenital malformation of the eye, chiefly characterized by iris hypoplasia, which can cause blindness. Pax-6 is involved in other anterior segment malformations besides aniridia, such as Peters anomaly, a major error in the embryonic development of the eye with corneal clouding with variable iridolenticulocorneal adhesions. The Pax-6 gene encodes a transcriptional regulator that recognizes target genes through its paired-type DNA-binding domain. The paired domain is composed of two distinct DNA-binding subdomains, the amino-terminal subdomain and the carboxy-terminal subdomain, which bind respective consensus DNA sequences. The human Pax-6 gene produces two alternatively spliced isoforms that have the distinct structure of the paired domain. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction