Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax6 Antibody (PAX6/7078R), DyLight 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314033UV
This item is not returnable.
View return policy
Description
Pax6 Monoclonal antibody specifically detects Pax6 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
| Pax6 | |
| Monoclonal | |
| DyLight 350 | |
| keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6 | |
| Recombinant fragment (N-terminus; aa 1-300) of human Pax6 protein (Uniprot: P26367) | |
| 0.1 mL | |
| Cellular Markers, Diabetes Research, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
| 5080 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| PAX6/7078R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction