Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax6 Antibody (PAX6/7078R), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314033MFV500
Description
Pax6 Monoclonal antibody specifically detects Pax6 in Human samples. It is validated for Flow CytometrySpecifications
Pax6 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
PAX6/7078R | |
Flow Cytometry | |
keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6 | |
Recombinant fragment (N-terminus; aa 1-300) of human Pax6 protein (Uniprot: P26367) | |
0.1 mL | |
Cellular Markers, Diabetes Research, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
5080 | |
Store at 4C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction