Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAX6 Monoclonal Antibody (7H10.D3.D4.G5), Rockland™
SDP

Catalog No. 501990672
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0672 100 μg
50-199-0673 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0672 Supplier Rockland Immunochemicals Supplier No. 200301R22
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Cross-reactivity with PAX6 from other sources has not been determined.

This gene encodes paired box gene 6, one of many human homologs of the Drosophila melanogaster gene prd. In addition to the hallmark feature of this gene family, a conserved paired box domain, the encoded protein also contains a homeo box domain. Both domains are known to bind DNA, and function as regulators of gene transcription. This gene is expressed in the developing nervous system, and in developing eyes. Mutations in this gene are known to cause ocular disorders such as aniridia and Peter's anomaly. Alternatively spliced transcript variants encoding either the same or different isoform have been found for this gene.

Specifications

Antigen PAX6
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 7H10.D3.D4.G5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene PAX6
Gene Accession No. P26367
Gene Alias 1500038E17Rik; AEY11; AN; AN2; Aniridia; Aniridia type II protein; D11S812E; Dey; Dickie's small eye; FVH1; Gsfaey11; Keratitis; MGC17209; MGDA; Oculorhombin; paired box 6; paired box gene 6; Paired Box Gene-6; paired box homeotic gene 6; paired box homeotic gene-6; Paired box protein Pax-6; Pax6; Pax-6; Sey; small eye; WAGR
Gene Symbols PAX6
Host Species Mouse
Immunogen PAX6 protein A purified antibody was prepared in mice by repeated immunizations with a recombinant protein corresponding to human PAX6 protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5080
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.