Learn More
PAX6 Monoclonal Antibody (E5B), Abbomax
Mouse Monoclonal Antibody
Supplier: Abbomax Inc 604250

Description
Positive control: Developing eyes Cellular location: Nucleus.
PAX6 Monoclonal antibody specifically detects PAX6 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
PAX6 | |
Monoclonal | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
P26367 | |
Mouse | |
Protein A | |
RUO | |
5080 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
E5B | |
Unconjugated | |
PAX6 | |
1500038E17Rik; AEY11; AN; AN2; Aniridia; aniridia type II protein; D11S812E; Dey; Dickie's small eye; FVH1; Gsfaey11; Keratitis; MGC17209; MGDA; oculorhombin; paired box 6; paired box gene 6; Paired Box Gene-6; paired box homeotic gene 6; paired box homeotic gene-6; paired box protein Pax-6; Pax6; Pax-6; PAX6 protein; Sey; small eye; transcription factor; WAGR | |
A synthetic peptide derived from human PAX6 protein | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.