Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAX6 Polyclonal Antibody, Bioss
SDP

Catalog No. 502281795
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1795 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-1795 Supplier Bioss Supplier No. BS22254R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PAX6 Polyclonal antibody specifically detects PAX6 in Human, Mouse, Rat samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PAX6
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene PAX6
Gene Accession No. P26367, P63015, P63016
Gene Alias 1500038E17Rik; AEY11; AN; AN2; Aniridia; aniridia type II protein; D11S812E; Dey; Dickie's small eye; FVH1; Gsfaey11; Keratitis; MGC17209; MGDA; oculorhombin; paired box 6; paired box gene 6; Paired Box Gene-6; paired box homeotic gene 6; paired box homeotic gene-6; paired box protein Pax-6; Pax6; Pax-6; PAX6 protein; Sey; small eye; transcription factor; WAGR
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PAX6 (221-320).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18508, 25509, 5080
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.