Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax6 Rabbit anti-Human, Clone: SD08-31, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267823
Description
Pax6 Monoclonal specifically detects Pax6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Pax6 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6 | |
Rabbit | |
Protein A purified | |
Cellular Markers, Diabetes Research, Neuronal Stem Cell Markers, Neuronal Stem Cells, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
5080 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
SD08-31 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
PAX6 | |
Recombinant protein within Human Pax6 aa 152-351/ 422. (SwissProt: P26367 Human; SwissProt: P63015 Mouse; SwissProt: P63016 Rat) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction