Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax7 Antibody (PAX7/1187) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$670.50
Specifications
Antigen | Pax7 |
---|---|
Clone | PAX7/1187 |
Concentration | 1.0 mg/mL |
Dilution | Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP247922A
![]() |
Novus Biologicals
NBP2479220.1MG |
0.1 mg |
Each for $670.50
|
|
|||||
NBP247922B
![]() |
Novus Biologicals
NBP2479220.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Pax7 Monoclonal specifically detects Pax7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Pax7 | |
1.0 mg/mL | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Apoptosis, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
PBS with No Preservative | |
5081 | |
Recombinant fragment (aa300-600) of human PAX7 protein | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
57 kDa |
PAX7/1187 | |
Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
FLJ37460, HuP1, paired box 7, paired box gene 7, paired box homeotic gene 7, paired box protein Pax-7, paired domain gene 7, PAX7 transcriptional factor, PAX7B, RMS2 | |
PAX7 | |
IgG1 κ | |
Protein A or G purified | |
The Pax gene family of nuclear transcription factors is comprised of nine members that function during embryogenesis to regulate the temporal and position-dependent differentiation of cells. In addition, the family is involved in a variety of signal transduction pathways in the adult organism. Mutations in the Pax family of proteins have been linked to disease and cancer in humans. Pax-7 is a protein specifically expressed in cultured satellite cell-derived myoblasts. In situ hybridization reveals that Pax-7 is also expressed in satellite cells residing in adult muscle. A chromosomal aberration in the gene encoding Pax-7 causes rhabdomyosarcoma 2 (RMS2) (also called alveolar rhabdomyosarcoma). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title