Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAX7 Monoclonal Antibody (2F12H4), Active Motif™

Rat Monoclonal Antibody
$156.65 - $620.58
Specifications
Antigen | PAX7 |
---|---|
Clone | 2F12H4 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2868
![]() ![]() |
Active Motif
39804 |
10 μg |
Each for $156.65
|
|
|||||
50-199-2867
![]() ![]() |
Active Motif
39803 |
100 μg |
Each for $620.58
|
|
|||||
Description
This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The specific function of the paired box 7 gene is unknown but speculated to involve tumor suppression since fusion of this gene with a forkhead domain family member has been associated with alveolar rhabdomyosarcoma. Three transcript variants encoding different isoforms have been found for this gene.Specifications
PAX7 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 | |
FLJ37460; HUP1; paired box 7; paired box gene 7; paired box homeotic gene 7; Paired box protein Pax-7; paired box transcription factor PAX7; paired domain gene 7; Pax7; Pax-7; PAX7 transcriptional factor; PAX7A; PAX7B; RGD1564360; RMS2 | |
PAX7 | |
IgG2b | |
Protein G | |
Antibody |
2F12H4 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
P47239 | |
18509 | |
This PAX7 antibody was raised against a peptide derived from mouse PAX7. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
PAX7 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title