Learn More
PAX7 Mouse anti-Human, Clone: SPM613, Abnova™
Mouse monoclonal antibody raised against partial recombinant human PAX7.
Supplier: Abnova Corporation MAB13136
Description
This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The specific function of the paired box 7 gene is unknown but speculated to involve tumor suppression since fusion of this gene with a forkhead domain family member has been associated with alveolar rhabdomyosarcoma. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
PAX7 | |
Monoclonal | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
HUP1/PAX7B | |
Mouse | |
Protein A/G purification | |
RUO | |
5081 | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Immunohistochemistry (PFA fixed) | |
SPM613 | |
Mouse monoclonal antibody raised against partial recombinant human PAX7. | |
In 10mM PBS. | |
PAX7 | |
Recombinant protein corresponding to amino acids 300-600 of human PAX7. | |
100 μg | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.