Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAX7 Rabbit anti-Human, Unconjugated, Clone: HGH-16, Abnova™

Catalog No. 89336695 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-336-695 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-336-695 Supplier Abnova Corporation Supplier No. MAB20295
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

PAX7 Monoclonal antibody specifically detects PAX7 in Human samples. It is validated for WB-Ce, Flow Cyt.

Specifications

Antigen PAX7
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone HGH-16
Conjugate Unconjugated
Description paired box 7
Gene PAX7
Gene Alias HUP1, PAX7B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human PAX7
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 5081
Target Species Human
Content And Storage Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.