Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAX8 Antibody (PAX8/1491 + PAX8/1492), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308274MFV610
Description
PAX8 Monoclonal antibody specifically detects PAX8 in Human samples. It is validated for Flow CytometrySpecifications
PAX8 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
PAX8/1491 + PAX8/1492 | |
Flow Cytometry | |
paired box 8, paired box gene 8, paired box protein Pax-8, paired domain gene 8 | |
Recombinant human PAX8 protein (Uniprot: Q06710) | |
0.1 mL | |
Cancer | |
7849 | |
Store at 4C in the dark. | |
IgG2a κ, IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction