Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAX8 Antibody (PAX8/2774R), DyLight 650, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308275C
Description
PAX8 Monoclonal antibody specifically detects PAX8 in Human, Canine samples. It is validated for Immunohistochemistry (Paraffin)Specifications
PAX8 | |
Monoclonal | |
DyLight 650 | |
paired box 8, paired box gene 8, paired box protein Pax-8, paired domain gene 8 | |
Recombinant fragment (around aa 60-261) of human PAX8 protein (exact sequence is proprietary) (Uniprot: Q06710) | |
100 μg | |
Cancer | |
7849 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PAX8/2774R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction