Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAX8 Monoclonal Antibody (2D4)
GREENER_CHOICE

Catalog No. PIMA515708
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515708 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515708 Supplier Invitrogen™ Supplier No. MA515708
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15708 targets PAX8 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15708 immunogen is purified recombinant fragment of human PAX8 expressed in E. Coli. MA5-15708 detects PAX8 which has a predicted molecular weight of approximately 60kDa.

This gene encodes a member of the paired box family of transcription factors. Members of this gene family typically encode proteins that contain a paired box domain, an octapeptide, and a paired-type homeodomain. This nuclear protein is involved in thyroid follicular cell development and expression of thyroid-specific genes. Mutations in this gene have been associated with thyroid dysgenesis, thyroid follicular carcinomas and atypical follicular thyroid adenomas. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAX8
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2D4
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene PAX8
Gene Accession No. Q06710
Gene Alias HGNC:8622; paired box 8; paired box gene 8; paired box protein Pax-8; paired domain gene 8; PAX8; Pax-8; Pax-8 protein
Gene Symbols PAX8
Host Species Mouse
Immunogen Purified recombinant fragment of human PAX8 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7849
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.