Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAX8 Antibody (PAX8/1491), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | PAX8 |
---|---|
Clone | PAX8/1491 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25326020
![]() |
Novus Biologicals
NBP25326020UG |
20 μg |
Each for $268.00
|
|
|||||
NB253260100
![]() |
Novus Biologicals
NBP253260100UG |
100 μg |
Each for $524.00
|
|
|||||
NB253260200
![]() |
Novus Biologicals
NBP253260200UG |
200 μg | N/A | N/A | N/A | ||||
Description
PAX8 Monoclonal specifically detects PAX8 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.Specifications
PAX8 | |
0.2 mg/mL | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cancer | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
7849 | |
Recombinant human PAX8 fragment (aa60-261) (exact sequence is proprietary) | |
Primary | |
Store at 4C. |
PAX8/1491 | |
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
paired box 8, paired box gene 8, paired box protein Pax-8, paired domain gene 8 | |
PAX8 | |
IgG2b | |
Protein A or G purified | |
62 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title