Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAX8 Polyclonal Antibody, Abbomax

Catalog No. 502204506
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4506 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4506 Supplier Abbomax Inc Supplier No. 620590
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Breast carcinoma Cellular location: Nucleus.

PAX8 Polyclonal antibody specifically detects PAX8 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PAX8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene PAX8
Gene Accession No. P51974, Q00288, Q06710
Gene Alias HGNC:8622; paired box 8; paired box gene 8; paired box protein Pax-8; paired domain gene 8; PAX8; Pax-8; Pax-8 protein
Host Species Rabbit
Immunogen A synthetic peptide derived from internal sequence of human PAX8
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18510, 7849, 81819
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.