Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAX8 Rabbit anti-Human, Unconjugated, Clone: PAX8/2774R, Abnova™

Catalog No. 89339672 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-672 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-672 Supplier Abnova Corporation Supplier No. RAB00657
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

PAX8 Monoclonal antibody specifically detects PAX8 in Human samples. It is validated for IHC-P.

Specifications

Antigen PAX8
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PAX8/2774R
Conjugate Unconjugated
Description paired box 8
Gene PAX8
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 60-261 of human PAX8
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7849
Target Species Human
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.