Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAX9 Monoclonal Antibody (7C2), Active Motif™
SDP

Catalog No. 501993116
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3116 100 μg
50-199-3117 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3116 Supplier Active Motif Supplier No. 61077
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The specific function of the paired box 9 gene is unknown but it may involve development of stratified squamous epithelia as well as various organs and skeletal elements.

Specifications

Antigen PAX9
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 7C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene pax9
Gene Accession No. P55771
Gene Alias BOS_20365; EMBL:BAE79272.1}; etID309776.17; paired box 9; paired box gene 9; paired box homeotic gene 9; Paired box protein Pax-9; paired domain gene 9; PAX9; Pax-9; pax9 {ECO:0000312; pax9 protein; Pax9a; Pax9b; STHAG3
Gene Symbols pax9
Host Species Rat
Immunogen This PAX9 antibody was raised against a recombinant protein corresponding to amino acids to 263-342 of mouse PAX9 protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5083
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.