Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAX9 Rabbit anti-Human, Unconjugated, Clone: HFE-16, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20297
Description
PAX9 Monoclonal antibody specifically detects PAX9 in Human samples. It is validated for WB-Ce.Specifications
PAX9 | |
Monoclonal | |
Unconjugated | |
PAX9 | |
A synthetic peptide corresponding to human PAX9 | |
Primary | |
Human | |
Liquid |
Western Blot | |
HFE-16 | |
paired box 9 | |
Rabbit | |
100 μg | |
5083 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction