Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAXBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113848 Supplier Invitrogen™ Supplier No. PA5113848
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGEIPDAAFI HAARKKRQMA RELGDFTPHD NEPGKGRLVR EDENDASDDE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

This gene encodes a protein that may bind to GC-rich DNA sequences, which suggests its involvement in the regulation of transcription. Alternative splicing of this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAXBP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PAXBP1
Gene Accession No. Q9Y5B6
Gene Alias 1810007M14Rik; BM020; C21orf66; FSAP105; functional spliceosome-associated protein 105; Gcfc; GCFC1; GC-rich sequence DNA binding factor 1; GC-rich sequence DNA-binding factor 1; GC-rich sequence DNA-binding factor candidate; GC-rich sequence DNA-binding factor homolog; PAX3 and PAX7 binding protein 1; PAX3- and PAX7-binding protein 1; PAX3/7BP; Paxbp1
Gene Symbols PAXBP1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human C21orf66 (aa 216-265).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94104
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.