Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAXBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552002
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552002 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552002 Supplier Invitrogen™ Supplier No. PA552002
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALSSLNVLRP GEIPDAAFIH AARKKRQMAR ELGDFTPHDN EPGKGRLVRE DENDASDDED DDEKRRIVFS VKEKSQRQKI AEEIGIEGSD DDALVTGEQD EELSRWEQEQ IRKGINIPQV QASQPAEVNM YYQNT Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.

This gene encodes a protein that may bind to GC-rich DNA sequences, which suggests its involvement in the regulation of transcription. Alternative splicing of this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAXBP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PAXBP1
Gene Accession No. Q9Y5B6
Gene Alias 1810007M14Rik; BM020; C21orf66; FSAP105; functional spliceosome-associated protein 105; Gcfc; GCFC1; GC-rich sequence DNA binding factor 1; GC-rich sequence DNA-binding factor 1; GC-rich sequence DNA-binding factor candidate; GC-rich sequence DNA-binding factor homolog; PAX3 and PAX7 binding protein 1; PAX3- and PAX7-binding protein 1; PAX3/7BP; Paxbp1
Gene Symbols PAXBP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PAXBP1. Recombinant protein control fragment (Product #RP-100751).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94104
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.